Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TdT Antibody (4B10C3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261898
Description
TdT Monoclonal specifically detects TdT in Human samples. It is validated for Western Blot, ELISA.Specifications
TdT | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, ELISA 1:10000 | |
deoxynucleotidyltransferase, terminal, EC 2.7.7, EC 2.7.7.31, nucleosidetriphosphate:DNA deoxynucleotidylexotransferase, TDTDNA nucleotidylexotransferase, Terminal addition enzyme, Terminal deoxynucleotidyltransferase, terminal deoxyribonucleotidyltransferase, Terminal transferase | |
Mouse | |
58.5 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
4B10C3 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
DNTT | |
Purified recombinant fragment of human DNTT (AA: 52-192) expressed in E. Coli. | |
Protein G purified | |
Adaptive Immunity, Cell Cycle and Replication, Immunology | |
1791 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction