Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TdT Antibody (DNTT/4506R), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB379829 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379829 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379829 Supplier Novus Biologicals Supplier No. NBP308564FR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TdT Monoclonal antibody specifically detects TdT in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen TdT
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DNTT/4506R
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias deoxynucleotidyltransferase, terminal, EC 2.7.7, EC 2.7.7.31, nucleosidetriphosphate:DNA deoxynucleotidylexotransferase, TDTDNA nucleotidylexotransferase, Terminal addition enzyme, Terminal deoxynucleotidyltransferase, terminal deoxyribonucleotidyltransferase, Terminal transferase
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues within aa1-100 of TdT was used as an immunogen. (Uniprot: P04053)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Cycle and Replication, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1791
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.