Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TdT Antibody (rDNTT/6909), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB156472 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB156472 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB156472 Supplier Novus Biologicals Supplier No. NBP314198AF405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TdT Monoclonal antibody specifically detects TdT in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen TdT
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rDNTT/6909
Conjugate Alexa Fluor 405
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias deoxynucleotidyltransferase, terminal, EC 2.7.7, EC 2.7.7.31, nucleosidetriphosphate:DNA deoxynucleotidylexotransferase, TDTDNA nucleotidylexotransferase, Terminal addition enzyme, Terminal deoxynucleotidyltransferase, terminal deoxyribonucleotidyltransferase, Terminal transferase
Host Species Mouse
Immunogen Recombinant fragment (around aa1-100) of human TdT protein (exact sequence is proprietary) (Uniprot: P04053)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Cycle and Replication, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1791
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.