Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TdT Monoclonal Antibody (7UNA8U6), eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

$139.50 - $346.50

Specifications

Antigen TdT
Clone 7UNA8U6
Concentration 0.5 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-255-0878
Encompass_Preferred
View Documents
Invitrogen™
14973980
25 μg
Each for $139.50

Save 25%

Reg : $186.00

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Edge
Add to Cart
 
14-973-982
Encompass_Preferred
View Documents
Invitrogen™
14973982
100 μg
Each for $346.50

Save 25%

Reg : $462.00

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Edge
Add to Cart
 
Description

Description

The MAb 7UNA8U6 reacts with human, mouse, and rat TdT. TdT, or terminal-deoxynucleotidyl transferase, is a 60 kDa polymerase responsible for the template-independent addition of N-nucleotides at gene segment junctions of developing lymphocytes. Expression is found in the nucleus of immature lymphocytes but not in mature lymphocytes or non-lymphoid cells. TdT plays a critical role during TCR and Ig gene rearrangement and functions to increase the diversity of the lymphocyte repertoire through the addition of N-nucleotides onto the ends of newly formed P-nucleotides. After positive selection, expression of TdT becomes decreased or absent. Expression of TdT has been shown to be present in lymphoma or lymphoblastic leukemia cells.

Terminal Deoxynucleotidyl Transferase (TdT) is a DNA polymerase located in the cell nucleus which catalyses the polymerization of deoxynucleotides at the 3' hydroxyl ends of oligo or polydeoxynucleotide initiators and functions without a template. TdT is considered to be a highly specific marker for the diagnosis and classification of acute lymphoblastic lymphoma/leuksemias. The determination of TdT expression is most valuable when it is different to differentiate histologically between lymphoblastic lymphoma and Burkitt's lymphoma.
Specifications

Specifications

TdT
0.5 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
Deoxynucleotidyltransferase terminal; deoxynucleotidyltransferase, terminal; DNA nucleotidylexotransferase; Dntt; Nucleosidetriphosphate DNA Deoxynucleotidylexotransferase; nucleosidetriphosphate:DNA deoxynucleotidylexotransferase; TDT; Terminal addition enzyme; terminal deoxynucleotidyl transferase; terminal deoxynucleotidyltransferase; terminal deoxyribonucleotidyltransferase; terminal transferase
DNTT
Primary
4°C
DNTT
7UNA8U6
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human, Mouse, Rat
P04053, P09838
1791, 21673, 294051
IgG1 κ
Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.