Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TdT Mouse anti-Human, Clone: TDT/1393, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP30767320UG

 View more versions of this product

Catalog No. NB122820


Only null left
Add to Cart

Description

Description

TdT Monoclonal specifically detects TdT in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

TdT
Monoclonal
0.2 mg/ml
ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
deoxynucleotidyltransferase, terminal, EC 2.7.7, EC 2.7.7.31, nucleosidetriphosphate:DNA deoxynucleotidylexotransferase, TDTDNA nucleotidylexotransferase, Terminal addition enzyme, Terminal deoxynucleotidyltransferase, terminal deoxyribonucleotidyltransferase, Terminal transferase
Recombinant fragment (around aa 52-192) of human TdT protein (exact sequence is proprietary)
20 μg
Adaptive Immunity, Cell Cycle and Replication, Immunology
1791
Store at 4°C.
IgG1 κ
ELISA, Immunohistochemistry (Paraffin), Peptide Array
TDT/1393
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.