Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TdT Mouse anti-Human, Clone: TDT/1393, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | TdT |
---|---|
Clone | TDT/1393 |
Concentration | 0.2 mg/ml |
Dilution | ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | ELISA, Immunohistochemistry (Paraffin), Peptide Array |
Description
TdT Monoclonal specifically detects TdT in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Protein Array.Specifications
TdT | |
0.2 mg/ml | |
ELISA, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Cell Cycle and Replication, Immunology | |
10 mM PBS with 0.05% BSA | |
1791 | |
IgG1 κ | |
Protein A or G purified |
TDT/1393 | |
ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
deoxynucleotidyltransferase, terminal, EC 2.7.7, EC 2.7.7.31, nucleosidetriphosphate:DNA deoxynucleotidylexotransferase, TDTDNA nucleotidylexotransferase, Terminal addition enzyme, Terminal deoxynucleotidyltransferase, terminal deoxyribonucleotidyltransferase, Terminal transferase | |
Recombinant fragment (around aa 52-192) of human TdT protein (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title