Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ TDZD 8

Click to view available options
Quantity:
10 mg
50 mg
Description
TDZD 8 is a selective non-ATP competitive inhibitor of GSK 3β (IC50 = 2 μM); thiadiazolidinone deriviative. Does not inhibit Cdk-1/cyclin B, CK-II, PKA or PKC at >100 μM. Reduces severity of L-dopa-induced dyskinesia in a Parkinson's disease in vivo model.
Specifications
Specifications
| CAS | 327036-89-5 |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO, 100mM in ethanol |
| Molecular Formula | C10H10N2O2S |
| Formula Weight | 222.26g/mol |
| Color | White |
| Product Type | Inhibitor |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction