Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEAD4 Rabbit anti-Human, Clone: S09-9D9, Novus Biologicals™
SDP

Catalog No. NB165405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165405 100 μg
NB165406 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB165405 Supplier Novus Biologicals Supplier No. NBP315099100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TEAD4 Monoclonal antibody specifically detects TEAD4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen TEAD4
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S09-9D9
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias EFTR-2, hRTEF-1B, related transcription enhancer factor 1B, RTEF1RTEF-1, TCF13L1MGC9014, TEA domain family member 4TEF3, TEAD-4, TEF-3, TEFR-1, Transcription factor 13-like 1, Transcription factor RTEF-1, transcriptional enhancer factor 1-related, transcriptional enhancer factor 3, transcriptional enhancer factor TEF-3
Host Species Rabbit
Immunogen Recombinant protein of human TEAD4 (Uniprot#: Q15561)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7004
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.