Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Teashirt homolog 1 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB183951 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183951 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB183951 Supplier Novus Biologicals Supplier No. NBP282038UV
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Teashirt homolog 1 Polyclonal antibody specifically detects Teashirt homolog 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen Teashirt homolog 1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias Antigen NY-CO-33, NY-CO-33, SDCCAG33, Serologically defined colon cancer antigen 33teashirt family zinc finger 1, teashirt homolog 1, teashirt zinc finger homeobox 1, TSH1teashirt zinc finger 1
Host Species Rabbit
Immunogen Teashirt homolog 1 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human Teashirt homolog 1. The immunogen is located within amino acids 70 - 120 of Teashirt homolog 1. Amino Acid Squence: AHFKGSSSREEKEDPQ
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10194
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.