Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Teashirt homolog 1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB341397 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341397 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341397 Supplier Novus Biologicals Supplier No. NBP282038MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Teashirt homolog 1 Polyclonal antibody specifically detects Teashirt homolog 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Teashirt homolog 1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Antigen NY-CO-33, NY-CO-33, SDCCAG33, Serologically defined colon cancer antigen 33teashirt family zinc finger 1, teashirt homolog 1, teashirt zinc finger homeobox 1, TSH1teashirt zinc finger 1
Host Species Rabbit
Immunogen Teashirt homolog 1 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human Teashirt homolog 1. The immunogen is located within amino acids 70 - 120 of Teashirt homolog 1. Amino Acid Squence: AHFKGSSSREEKEDPQ
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10194
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.