Description
- Variable width webbing in sub-pelvic area distributes weight over a wider area for increased comfort and support
- PTFE-coated webbing resists water, oil, grease and dirt
- Adjustable D-rings
- Qwik-Fit chest buckle
- Integral body belt with two side D-rings and two accessory rings and removable saddle with adjustable height
- Secure-Fit buckles feature single-hand operation, and positive engagement window
- Adjustable Y-back D-locator pad allows complete adjustment of back D-ring for proper fit
Certifications
CSA certified
Compliance
Meet applicable OSHA and ANSI standards.
Specifications
Specifications
| D-Ring Location | Back, Chest, Hips |
| Buckle Style | Secure-Fit |
| Size | Standard |
| Style | Vest |
| Certifications/Compliance | CSA certified; Meet applicable OSHA and ANSI standards. |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.