Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Techne™ PrimePRO QPCR DNA detection Kit, Neisseria gonorrhoeae (with Mastermix)

Catalog No. TKIT10119M Shop All Oakton Products
Change view
Click to view available options
150 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
TKIT10119M 150 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TKIT10119M Supplier Techne™ Supplier No. 9996768

missing translation for 'validMainframeMsg'

  • Kits packed in a convenient foil pouch
  • Lyophilized (freeze dried)
  • No dry ice shipping required
  • Reduced shipping costs
  • Simple protocol, identical for each kit
  • 150 reactions per kit
  • Low cost per test
  • qPCR can be days faster, more accurate and more quantifiable than traditional microbiological methods
  • Suitable for any qPCR instrument with FAM/VIC detection

Specifications

Content And Storage Lyophilized primer and probe mix, Lyophilized master mix, Lyophilized master mix resuspension buffer, Lyophilized ROX Ampule, High-copy DNA for standard curve, Internal extraction control DNA, Beta-actin endogenous control, RNAse/DNAse free water, Reverse transcriptase (RNA kits only)
Store at ambient temperature, stable for 18 months
Detection Method qPCR
For Use With (Equipment) any qPCR instrument with FAM/VIC detection
Kit Type QPCR DNA Detection Kit
No. of Tests 150
Quantity 150 Reactions
For Use With (Application) Microbial Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.