Learn More
Techne™ PrimePRO QPCR DNA detection Kit, Staphylococcus aureus (without Mastermix)
Shop All Oakton ProductsDescription
- Kits packed in a convenient foil pouch
- Lyophilized (freeze dried)
- No dry ice shipping required
- Reduced shipping costs
- Simple protocol, identical for each kit
- 150 reactions per kit
- Low cost per test
- qPCR can be days faster, more accurate and more quantifiable than traditional microbiological methods
- Suitable for any qPCR instrument with FAM/VIC detection
Specifications
Specifications
| Content And Storage | Lyophilized primer and probe mix, Lyophilized master mix, Lyophilized master mix resuspension buffer, Lyophilized ROX Ampule, High-copy DNA for standard curve, Internal extraction control DNA, Beta-actin endogenous control, RNAse/DNAse free water Store at ambient temperature, stable for 18 months |
| Detection Method | qPCR |
| For Use With (Equipment) | any qPCR instrument with FAM/VIC detection |
| Kit Type | QPCR DNA Detection Kit |
| No. of Tests | 150 |
| Quantity | 150 Reactions |
| For Use With (Application) | Microbial Detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.