Learn More
Johannes Lieder Wood Sections (Transverse, Radial, and Tangential)
Shop All Johannes Lieder GmbH & Co. KG Products
Click to view available options
Mounting Type:
Cross and Longitudinal Section
Description
- 7200 Wood Sections. Each slide comprises three sections: transverse,radial and tangential section.
- 25 Microscope Slides
- Maple. Acer platanoides
- Apple. Pyrus malus
- Birch. Betula pendula
- Pear. Pyrus communis
- Mountain ash. Sorbus aucuparia
- Yew. Taxus baccata
- Oak. Quercus robur
- Alder. Alnus glutinosa
- Ash. Fraxinus excelsior
- Spruce. Picea excelsa
- White beech. Carpinus betulus
- Pine. Pinus silvestris
- Cherry. Prunus avium
- Larch. Larix decidua
- Lime. Tilia platyphylla
- Walnut. Juglans regia
- Poplar. Populus alba
- Plane. Platanus orientalis
- Plum. Prunus domestica
- Black locust. Robinia pseudacacia
- Chestnut. Aesculus hippocastanum
- Beech. Fagus silvatica
- Elm. Ulmus scabra
- Willow. Salix alba
- Fir. Abies alba
Specifications
Specifications
| Classification | General |
| Material | Elastin Stained Glass |
| Mounting Type | Cross and Longitudinal Section |
| Type | Microscope Slides |
| Slide Type | Prepared Slide |