Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEDC1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB319759 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB319759 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB319759 Supplier Novus Biologicals Supplier No. NBP298663JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TEDC1 Polyclonal antibody specifically detects TEDC1 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen TEDC1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias C14orf80, chromosome 14 open reading frame 80
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TEDC1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283643
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.