Learn More
MilliporeSigma™ Supelco™ Tedlar™ PLV Gas Sampling Bag with Thermogreen™ LB-2 Septa
Tedlar™ is recommended in several US EPA methods for sampling VOCs and sulfur compounds.
Supplier: MilliporeSigma™ Supelco™ 30291U
Description
Tedlar™ is made from a high-quality 2 mil Tedlar™ film for superior inertness, impermeability, sample integrity and good recovery data. Our Push Lock Valve (PLV) and seam-sealing process ensure that these bags are sturdy and virtually leak-proof, even under the most demanding sampling and shipping conditions.
Specifications
| 3 L | |
| PVDF Film | |
| 10/Pk. |
| Push/Pull Lock Valve (PLV) | |
| Tedlar |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tedlar is a registered trademark of E. I. du Pont de Nemours and Company. Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC.
Spot an opportunity for improvement?Share a Content Correction