Learn More
MilliporeSigma™ Supelco™ Tedlar™ PLV Gas Sampling Bag with Thermogreen™ LB-2 Septa
Tedlar™ is recommended in several US EPA methods for sampling VOCs and sulfur compounds.
Supplier:  MilliporeSigma™ Supelco™ 24654
 View more versions of this product
Catalog No. 11-102-0817
Only null left
Add to Cart
Description
Description
Tedlar™ is made from a high-quality 2 mil Tedlar™ film for superior inertness, impermeability, sample integrity and good recovery data. Our Push Lock Valve (PLV) and seam-sealing process ensure that these bags are sturdy and virtually leak-proof, even under the most demanding sampling and shipping conditions.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 L
PVDF Film
10/Pk.
Push/Pull Lock Valve (PLV)
Tedlar
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
Tedlar is a registered trademark of E. I. du Pont de Nemours and Company. Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC.