Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEM5/GPR124 Antibody (499716), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB310649 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310649 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310649 Supplier Novus Biologicals Supplier No. FAB8896PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TEM5/GPR124 Monoclonal antibody specifically detects TEM5/GPR124 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen TEM5/GPR124
Applications Flow Cytometry
Classification Monoclonal
Clone 499716
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias DKFZp434C211, DKFZp434J0911, G protein-coupled receptor 124, G-protein coupled receptor 124, KIAA1531FLJ14390, TEM5Tumor endothelial marker 5
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human TEM5/GPR124, Met1-Leu699, Accession # Q96PE1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endothelial Cell Markers, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 25960
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.