Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEM5/GPR124 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176844 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176844 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176844 Supplier Novus Biologicals Supplier No. NBP176844
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TEM5/GPR124 Polyclonal specifically detects TEM5/GPR124 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TEM5/GPR124
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 - 4 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 5 ug/ml
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q96PE1
Gene Alias DKFZp434C211, DKFZp434J0911, G protein-coupled receptor 124, G-protein coupled receptor 124, KIAA1531FLJ14390, TEM5Tumor endothelial marker 5
Gene Symbols ADGRA2
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide near the center of human TEM5. The immunogen is located within amino acids 560 - 610 of TEM5. Amino Acid Squence: PPEPEPPADQQLR
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Endothelial Cell Markers, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 25960
Target Species Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.