Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEM7/PLXDC1 Antibody (197C193 (IM193)), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP233228A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP233228A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP233228A70 Supplier Novus Biologicals Supplier No. NBP233228AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TEM7/PLXDC1 Monoclonal specifically detects TEM7/PLXDC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen TEM7/PLXDC1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 197C193 (IM193)
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FLJ45632, plexin domain containing 1,2410003I07Rik, plexin domain-containing protein 1, TEM3FLJ36270, TEM7DKFZp686F0937, Tumor endothelial marker 3, Tumor endothelial marker 7
Gene Symbols PLXDC1
Host Species Mouse
Immunogen Amino acids 409-425 (LQNNLSPKTKGTPVHLG) of human TEM7 were used to develop this monoclonal antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 57125.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.