Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEM7/PLXDC1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB323350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB323350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB323350 Supplier Novus Biologicals Supplier No. NBP299869PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TEM7/PLXDC1 Polyclonal antibody specifically detects TEM7/PLXDC1 in Human samples. It is validated for ELISA

Specifications

Antigen TEM7/PLXDC1
Applications ELISA
Classification Polyclonal
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias FLJ45632, plexin domain containing 1,2410003I07Rik, plexin domain-containing protein 1, TEM3FLJ36270, TEM7DKFZp686F0937, Tumor endothelial marker 3, Tumor endothelial marker 7
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human TEM7/PLXDC1 (Accession#: AAH36059.1; Met1-Thr426)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 57125
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.