Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEM8/ANTXR1 Antibody (200C1339(SB20)), Biotin, Novus Biologicals™
SDP

Catalog No. NB10056585B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056585B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056585B Supplier Novus Biologicals Supplier No. NB10056585B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TEM8/ANTXR1 Monoclonal specifically detects TEM8/ANTXR1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen TEM8/ANTXR1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 200C1339(SB20)
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Accession No. Q9H6X2
Gene Alias anthrax toxin receptor 1, ATR2310008J16Rik, FLJ10601, FLJ11298, FLJ21776, TEM82810405N18Rik, Tumor endothelial marker 8
Gene Symbols ANTXR1
Host Species Mouse
Immunogen This monoclonal antibody was developed by immunizing mice with a synthetic peptide containing amino acids 481-497 (CINFTRVKNNQPAKYPL) of human TEM8.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 84168
Test Specificity Nanda et al (2004) note a passage dependent loss of TEM8 in HUVEC suggesting that that TEM8 may be a specific marker of the endothelial cell (EC) phenotype which is lost as primary EC's age in culture and de-differentiate.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.