Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEM8/ANTXR1 Antibody, PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB214068 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB214068 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB214068 Supplier Novus Biologicals Supplier No. NBP177233PECY7
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TEM8/ANTXR1 Polyclonal antibody specifically detects TEM8/ANTXR1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry

Specifications

Antigen TEM8/ANTXR1
Applications Flow Cytometry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate PE-Cyanine7
Formulation PBS
Gene Alias anthrax toxin receptor 1, ATR2310008J16Rik, FLJ10601, FLJ11298, FLJ21776, TEM82810405N18Rik, Tumor endothelial marker 8
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 13 amino acids near the center of human ATR. The immunogen is located within amino acids 220 - 270 of ATR. Amino Acid Squence: RGNGFRHARNVDR
Purification Method Ion exchange chromatography
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 84168
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.