Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEM8/ANTXR1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB340408 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340408 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340408 Supplier Novus Biologicals Supplier No. NBP321501PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TEM8/ANTXR1 Polyclonal antibody specifically detects TEM8/ANTXR1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen TEM8/ANTXR1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias anthrax toxin receptor 1, ATR2310008J16Rik, FLJ10601, FLJ11298, FLJ21776, TEM82810405N18Rik, Tumor endothelial marker 8
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 13 amino acids near the C-terminus of human TEM8/ANTXR1. The immunogen is located within the last 50 amino acids.. Amino Acid Squence: RAPPPSRPPPRPSV
Purification Method Ion exchange chromatography
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 84168
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.