Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Temsirolimus, 99+%

Catalog No. p-7082368
Click to view available options
Quantity:
25 mg
50 mg
100 mg

A Rapamycin derivative with anti-cancer activity that inhibits mTOR | CAS: 162635-04-3 | C56H87NO16

Temsirolimus is used as a rapamycin derivative with anti-cancer activity that inhibits mTOR. Temsirolimus is an antiproliferative and antiangiogenic, the first-in-class mTOR inhibitor approved for the treatment of patients with advanced poor prognosis renal cell carcinoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162635-04-3
Molecular Formula C56H87NO16

Specifications

Quantity 100 mg
Description White to Off-white
Content And Storage -30°C to -10°C
For Use With (Application) A Rapamycin derivative with anti-cancer activity that inhibits mTOR
Formula Weight 1030.29g/mol
Product Type Signal Transduction Reagent
Form Powder
Assay ≥99%
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.