Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tenascin C Antibody (TNC/2981R), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB389153 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389153 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389153 Supplier Novus Biologicals Supplier No. NBP308946AF750
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Tenascin C Monoclonal antibody specifically detects Tenascin C in Human, Rat (Negative) samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Tenascin C
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TNC/2981R
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias 150-225, Cytotactin, Glioma-associated-extracellular matrix antigen, GMEM, GP 150-225, Hexabrachion, hexabrachion (tenascin C, cytotactin), hexabrachion (tenascin), HXBcytotactin, JI, MGC167029, Myotendinous antigen, neuronectin, tenascin, tenascin C, Tenascin-C, tenascin-C isoform 14/AD1/16, TN-C, TNGP
Host Species Rabbit
Immunogen Recombinant human full-length Tenascin C protein (Uniprot: P24821)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Extracellular Matrix, Neuroscience, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3371
Target Species Human, Rat (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.