Learn More
Description
Specifications
Specifications
| Antigen | Tenascin X |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Hexabrachion-like protein, HXBL, tenascin XB, tenascin XB2, tenascin-X, TNXB1growth-inhibiting protein 45, TNXB2tenascin XB1, TNXBSTNXTENX, XBS, XBTN-X |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Tenascin X. |
| Purification Method | Antigen Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
