Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tenidap, Tocris Bioscience™

Catalog No. 258010
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 25-801-0

Supplier: R&D Systems 2580/10

This item is not returnable. View return policy

NSAID, cyclooxygenase (COX-1) inhibitor. Also opener of KIR2.3

NSAID that preferentially inhibits COX-1 (IC50 values are < 0.03, 1.2 and > 30 μM for COX-1, COX-2 and 5-lipoxygenase respectively). Inhibits formation of pro-inflammatory arachidonic acid metabolites in isolated human peripheral polymorphonuclear leukocytes. Opener of inward rectifying hKir2.3 channel (EC50 = 402 nM).

Chemical Identifiers

CAS 120210-48-2
Molecular Formula C14H9ClN2O3S
Molecular Weight (g/mol) 320.747
InChI Key LXIKEPCNDFVJKC-VAWYXSNFSA-N
Synonym tenidap sodium salt, dsstox_cid_26104, dsstox_rid_81342, dsstox_gsid_46104, e-5-chloro-3-hydroxy thiophen-2-yl methylene-2-oxoindoline-1-carboxamide, e-tenidap, 5-chloro-2-hydroxy-3-thiophene-2-carbonyl indole-1-carboxamide, 5-chloro-2-hydroxy-3-thiophen-2-ylcarbonyl-1h-indole-1-carboxamide, 5-chloro-2-hydroxy-3-thiophene-2-carbonyl-1h-indole-1-carboxamide, 5-chloro-2,3-dihydro-3-hydroxy 2-thienyl methylene-2-oxo-1h-indole-1-carboxamide
PubChem CID 54704052
IUPAC Name (3E)-5-chloro-3-[hydroxy(thiophen-2-yl)methylidene]-2-oxoindole-1-carboxamide
SMILES C1=CSC(=C1)C(=C2C3=C(C=CC(=C3)Cl)N(C2=O)C(=O)N)O

Specifications

Quantity 10 mg
Formula Weight 320.75
Percent Purity >99%
Chemical Name or Material Tenidap
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.