Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Teniposide 98.0+%, TCI America™
SDP

Catalog No. T3109100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20mg
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3109100MG 100mg
T310920MG 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3109100MG Supplier TCI America Supplier No. T3109100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29767-20-2
Molecular Formula C32H32O13S

Specifications

Quantity 100mg
Melting Point 247°C
Formula Weight 656.66
Physical Form Crystalline Powder at 20°C
Color White
Purity ≥98.0% (HPLC)
Chemical Name or Material Teniposide
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.