Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Tenofovir

Click to view available options
Quantity:
10 mg
50 mg
Description
Tenofovir is a selectively inhibits HIV reverse transcriptase (RNA-dependent DNA polymerase). Prevents cytotoxicity in SIV-infected C-8166 cells in vitro (IC50 = 1.5 μM). Antiviral agent.
Specifications
Specifications
CAS | 147127-20-6 |
Quantity | 50 mg |
Molecular Formula | (R)-[[2-(6-Amino-9 H-purin-9-yl)-1-methylethoxy]methyl]phosphonic acid |
Product Type | Tenofovir |
Purity | 99% |
Molecular Weight (g/mol) | M.W. (Observed): 287.21 |
Inhibitors | Tenofovir |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction