Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tensin 1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB336800 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB336800 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB336800 Supplier Novus Biologicals Supplier No. NBP278783MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tensin 1 Polyclonal antibody specifically detects Tensin 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Tensin 1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DKFZp586K0617, matrix-remodelling associated 6, Matrix-remodelling-associated protein 6, MGC88584, MST091, MST122, MST127, MSTP127, MXRA6, tensin, tensin 1, tensin-1, TNSMSTP122
Host Species Rabbit
Immunogen Tensin 1 antibody made to partial recombinant protein made to the N-terminal portion of human Tensin 1 (amino acids 1-385) [UniProt Q9HBL0].
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7145
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.