Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Teplizumab Antibody (2505B) [PerCP], Novus Biologicals™
SDP

Catalog No. NB402196 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402196 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402196 Supplier Novus Biologicals Supplier No. FAB10245C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Teplizumab Monoclonal antibody specifically detects Teplizumab in Broad Species Reactivity samples. It is validated for Western Blot, Flow Cytometry

Specifications

Antigen Teplizumab
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 2505B
Conjugate PerCP
Dilution Western Blot, Flow Cytometry
Formulation PBS
Host Species Rabbit
Immunogen Full-length Teplizumab
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Broad Species Reactivity
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.