Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Teprotumumab Antibody (975547), HRP, Novus Biologicals™
SDP

Catalog No. NB345647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345647 Supplier Novus Biologicals Supplier No. FAB96341H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Teprotumumab Monoclonal specifically detects Teprotumumab in Multi-species samples. It is validated for Binding Inhibition, Flow Cytometry.

Specifications

Antigen Teprotumumab
Applications Binding Inhibition Assay, Flow Cytometry
Classification Monoclonal
Clone 975547
Conjugate HRP
Dilution Flow Cytometry, Binding Inhibition
Formulation PBS
Host Species Mouse
Immunogen Full-length Teprotumumab
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Multi-species
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.