Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TER-119 Antibody (TER-119), PerCP, Novus Biologicals™
SDP

Catalog No. NB297444 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297444 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297444 Supplier Novus Biologicals Supplier No. FAB1125C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TER-119 Monoclonal antibody specifically detects TER-119 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF-reported

Specifications

Antigen TER-119
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF
Classification Monoclonal
Clone TER-119
Conjugate PerCP
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF-reported
Formulation PBS
Gene Alias Ly76, Ly-76, TER119
Host Species Rat
Immunogen C57BL/6 mouse day-14 fetal liver cells
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104231
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.