Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Terazosin hydrochloride
GREENER_CHOICESDP

Catalog No. 150650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-065-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-065-0 Supplier Tocris Bioscience™ Supplier No. 1506/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

α1 and α2B antagonist (α1 > α2B). Orally active

α1- and α2B-adrenoceptor antagonist (Ki values are 3.3, 0.7, 1.1, 7.7, 1510 and 78.2 nM for α1A, α1B, α1D, α2B, α2A and α2C receptors respectively). Antihypertensive following oral or intravenous administration in vivo.

Chemical Identifiers

CAS 63074-08-8
Molecular Formula C19H26ClN5O4

Specifications

Quantity 50 mg
Formula Weight 423.9
Purity >99%
Molecular Weight (g/mol) 423.898
Chemical Name or Material Terazosin hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.