Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog Number p-7049255
Click to view available options
Quantity:
2 g
10 g
50 g
10043-27-3
N3O9Tb
344.94
MFCD00011257
YJVUGDIORBKPLC-UHFFFAOYSA-N
terbium(3+) trinitrate
[Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
10043-27-3
N3O9Tb
344.94
MFCD00011257
YJVUGDIORBKPLC-UHFFFAOYSA-N
terbium(3+) trinitrate
[Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Terbium(III) nitrate hydrate is used in ceramics, glass, phosphors, lasers, and also is the important dopant for fibre amplifiers. It is also used in alloys and in the production of electronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Physical Form Crystalline Aggregates
Quantity 2 g
UN Number UN1477
Merck Index 14,9157
Solubility Information Soluble in water.
Formula Weight 344.94 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Assay (REO)
Chemical Name or Material Terbium(III) nitrate hydrate
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECSNumber 233-138-7
TSCA Yes
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit