Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TERF2IP Antibody (78B356.1), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB10056526Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056526Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056526Y Supplier Novus Biologicals Supplier No. NB10056526AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TERF2IP Monoclonal specifically detects TERF2IP in Human samples. It is validated for Western Blot.

Specifications

Antigen TERF2IP
Applications Western Blot
Classification Monoclonal
Clone 78B356.1
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q9NYB0
Gene Alias dopamine receptor interacting protein 5, Dopamine receptor-interacting protein 5, DRIP5, hRap1, RAP1 homolog, RAP1TERF2-interacting telomeric protein 1, Repressor/activator protein 1 homolog, telomeric repeat binding factor 2, interacting protein, telomeric repeat-binding factor 2-interacting protein 1, TRF2-interacting telomeric protein 1, TRF2-interacting telomeric RAP1 protein
Gene Symbols TERF2IP
Host Species Mouse
Immunogen A synthetic peptide corresponding amino acids 212-235 of human Rap1 was used to immunize mice.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, DNA Repair, Endosome Markers, Lysosome Markers
Primary or Secondary Primary
Gene ID (Entrez) 54386
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.