Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TERF2IP Antibody, Novus Biologicals™
SDP

Catalog No. NB10056321 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056321 0.1 mg
NB10056321T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056321 Supplier Novus Biologicals Supplier No. NB10056321
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

TERF2IP Polyclonal specifically detects TERF2IP in Gerbil, Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot.

Specifications

Antigen TERF2IP
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunoprecipitation
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q9NYB0
Gene Alias dopamine receptor interacting protein 5, Dopamine receptor-interacting protein 5, DRIP5, hRap1, RAP1 homolog, RAP1TERF2-interacting telomeric protein 1, Repressor/activator protein 1 homolog, telomeric repeat binding factor 2, interacting protein, telomeric repeat-binding factor 2-interacting protein 1, TRF2-interacting telomeric protein 1, TRF2-interacting telomeric RAP1 protein
Gene Symbols TERF2IP
Host Species Rabbit
Immunogen Synthetic peptides corresponding amino acids 212-235 of human Rap1 was used to immunize rabbits.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Chromatin Research, DNA Repair, Endosome Markers
Primary or Secondary Primary
Gene ID (Entrez) 54386
Test Specificity Cross reacts with Human. Mouse reactivity reported in scientific literature (PMID: 24797474).
Target Species Gerbil, Mouse, Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.