Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TERF2IP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 6 publications
$206.50 - $426.00
Specifications
Antigen | TERF2IP |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunoprecipitation |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056321T
![]() |
Novus Biologicals
NB10056321SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056321
![]() |
Novus Biologicals
NB10056321 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
TERF2IP Polyclonal specifically detects TERF2IP in Human, Mouse, Gerbil samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
TERF2IP | |
Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunoprecipitation | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Gerbil | |
Q9NYB0 | |
54386 | |
Synthetic peptides corresponding amino acids 212-235 of human Rap1 was used to immunize rabbits. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Chromatin Research, DNA Repair, Endosome Markers | |
PBS with 0.02% Sodium Azide | |
dopamine receptor interacting protein 5, Dopamine receptor-interacting protein 5, DRIP5, hRap1, RAP1 homolog, RAP1TERF2-interacting telomeric protein 1, Repressor/activator protein 1 homolog, telomeric repeat binding factor 2, interacting protein, telomeric repeat-binding factor 2-interacting protein 1, TRF2-interacting telomeric protein 1, TRF2-interacting telomeric RAP1 protein | |
TERF2IP | |
IgG | |
Protein G purified | |
Cross reacts with Human. Mouse reactivity reported in scientific literature (PMID: 24797474). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title