Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TERF2IP Antibody, Novus Biologicals™
SDP

Catalog No. NB100292 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100292 Supplier Novus Biologicals Supplier No. NB100292
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 18 publications

TERF2IP Polyclonal specifically detects TERF2IP in Human samples. It is validated for FISH, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen TERF2IP
Applications FISH, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, Immunohistochemistry 1:500 - 1:2:000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:250 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:2:000
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. Q9NYB0
Gene Alias dopamine receptor interacting protein 5, Dopamine receptor-interacting protein 5, DRIP5, hRap1, RAP1 homolog, RAP1TERF2-interacting telomeric protein 1, Repressor/activator protein 1 homolog, telomeric repeat binding factor 2, interacting protein, telomeric repeat-binding factor 2-interacting protein 1, TRF2-interacting telomeric protein 1, TRF2-interacting telomeric RAP1 protein
Gene Symbols TERF2IP
Host Species Rabbit
Immunogen Antibody was generated using recombinant full length human RAP1 (Telomeric repeat binding factor 2 interacting protein 1) according to entry NP_061848.2
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, DNA Repair, Endosome Markers, Lysosome Markers
Primary or Secondary Primary
Gene ID (Entrez) 54386
Test Specificity TERF21P- Lysosome Marker.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.