Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octyl 4-methoxycinnamate, 98%, stabilized

Catalog No. AC291161000
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 5466-77-3 | C18H26O3 | 290.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5466-77-3
Molecular Formula C18H26O3
Molecular Weight (g/mol) 290.40
MDL Number MFCD00072582
InChI Key YBGZDTIWKVFICR-UHFFFAOYNA-N
Synonym bidd:er0152, octyl methoxy cinnamate omc, unii-4y5p7mud51 component, 2s-2-ethylhexyl 2e-3-4-methoxyphenyl prop-2-enoate
PubChem CID 11044481
IUPAC Name [(2S)-2-ethylhexyl] (E)-3-(4-methoxyphenyl)prop-2-enoate
SMILES CCCCC(CC)COC(=O)C=CC1=CC=C(OC)C=C1

Specifications

Acidity 1mg KOH/g max.
Absorbance 830 (at 307 to 308nm in methanol) min.
Color Colorless to Yellow
Density 1.0090g/mL
Flash Point 193°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5430 to 1.5470
Quantity 100 g
Specific Gravity 1.009
Merck Index 15,6859
Stabilizer 0.05 to 0.1% BHT
Formula Weight 290.39
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Octyl 4-methoxycinnamate
Show More Show Less
EINECSNumber 226-775-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.