Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TERT Antibody (2D8), Novus Biologicals™
SDP

Catalog No. NB100297SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100297SS 0.025 mL
NB100297 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100297SS Supplier Novus Biologicals Supplier No. NB100297SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 9 publications

TERT Monoclonal specifically detects TERT in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Western Blot.

Specifications

Antigen TERT
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Western Blot
Classification Monoclonal
Clone 2D8
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:200, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:50-1:100, Knockdown Validated
Formulation Ascites with 0.1% Sodium Azide
Gene Accession No. O14746
Gene Alias EST2Telomerase-associated protein 2, hEST2, TCS1EC 2.7.7.49, Telomerase catalytic subunit, telomerase reverse transcriptase, TP2HEST2, TRTEC 2.7.7
Gene Symbols TERT
Host Species Mouse
Immunogen Full-length recombinant human Telomerase reverse transcriptase from insect cells. [UniProt# O14746]
Molecular Weight of Antigen 127 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Embryonic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7015
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.