Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TERT Antibody (2D8), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 9 publications

$192.94 - $529.95

Specifications

Antigen TERT
Clone 2D8
Dilution Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:200, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:50-1:100, Knockdown Validated
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB100297SS
SDP
View Documents
Novus Biologicals
NB100297SS
0.025 mL
Each for $192.94
Only null left
Add to Cart
 
NB100297
SDP
View Documents
Novus Biologicals
NB100297
0.1 mL
Each for $529.95
Only null left
Add to Cart
 
Description

Description

TERT Monoclonal specifically detects TERT in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.
Specifications

Specifications

TERT
Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:200, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:50-1:100, Knockdown Validated
Monoclonal
Ascites
RUO
Human
O14746
7015
Full-length recombinant human Telomerase reverse transcriptase from insect cells. [UniProt# O14746]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
2D8
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Autophagy, Cancer, Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Embryonic Stem Cell Markers, Stem Cell Markers
Ascites with 0.1% Sodium Azide
EST2Telomerase-associated protein 2, hEST2, TCS1EC 2.7.7.49, Telomerase catalytic subunit, telomerase reverse transcriptase, TP2HEST2, TRTEC 2.7.7
TERT
IgM
Unpurified
127 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.