Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TERT Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | TERT |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL |
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TERT Polyclonal specifically detects TERT in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
TERT | |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
EST2Telomerase-associated protein 2, hEST2, TCS1EC 2.7.7.49, Telomerase catalytic subunit, telomerase reverse transcriptase, TP2HEST2, TRTEC 2.7.7 | |
TERT | |
IgG | |
Affinity Purified |
Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL | |
Polyclonal | |
Rabbit | |
Autophagy, Cancer, Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Embryonic Stem Cell Markers, Mitophagy, Stem Cell Markers | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
7015 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GVLLKTHCPLRAAVTPAAGVCAREKPQGSVAAPEEEDTDPRRLVQLLRQHSSPWQVYGFVRACLRRLVPPGLWGSRHNERRFLRNTKKFISLGKHAKLS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title