Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl carbamate, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
4248-19-5
C5H11NO2
117.15
LFKDJXLFVYVEFG-UHFFFAOYSA-N
t-butyl carbamate,carbamic acid tert-butyl ester,boc amine,boc-amide,carbamic acid, 1,1-dimethylethyl ester,boc-nh2,tert-butylcarbamat,n-boc amine,boc-amine,tert butyl carbamate
77922
tert-butyl carbamate
CC(C)(C)OC(=O)N
4248-19-5
C5H11NO2
117.15
LFKDJXLFVYVEFG-UHFFFAOYSA-N
t-butyl carbamate,carbamic acid tert-butyl ester,boc amine,boc-amide,carbamic acid, 1,1-dimethylethyl ester,boc-nh2,tert-butylcarbamat,n-boc amine,boc-amine,tert butyl carbamate
77922
tert-butyl carbamate
CC(C)(C)OC(=O)N
Packaging Glass bottle
Color White to Yellow
Melting Point 105.0°C to 109.0°C
Infrared Spectrum Authentic
Quantity 1g
Assay Percent Range 97.5% min. (GC)
Linear Formula H2NCO2C(CH3)3
Beilstein 03,II,26
Solubility Information Solubility in water: insoluble
Formula Weight 117.15
Physical Form Needles
Percent Purity 98%
Chemical Name or Material tert-Butyl carbamate
Show More Show Less

Warning

EINECSNumber : 224-209-3