Learn More
tert-Butyl ethyl ether, 99%
Description
tert-Butyl ethyl ether is used as an oxygenate gasoline additive oxygenate during its production from crude oil. It is used as an extractant in human urine by using single-walled carbon nanotubes as an adsorbent. It plays an important role as a fuel component in petrol to enhance its octane rating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
tert-Butyl ethyl ether is used as an oxygenate gasoline additive oxygenate during its production from crude oil. It is used as an extractant in human urine by using single-walled carbon nanotubes as an adsorbent. It plays an important role as a fuel component in petrol to enhance its octane rating.
Solubility
Miscible with alcohol, ethyl ether. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents, neoprene, n-butyl and urethane. It may react with air to form peroxides.

Specifications
Specifications
Melting Point | -94°C |
Density | 0.75 |
Boiling Point | 72°C to 73°C |
Flash Point | −19°C (−2°F) |
Refractive Index | 1.375 |
Quantity | 25 g |
UN Number | UN1179 |
Beilstein | 1731469 |
Merck Index | 14,3774 |
Solubility Information | Miscible with alcohol,ethyl ether. Slightly miscible with water. |
Show More |
Safety and Handling
Hazard Category | H225-H336 |
Hazard Statement | GHS H Statement H225-H331-H315-H319-H335 Highly flammable liquid and vapour. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL BUTYL ETHER |
EINECSNumber | 211-309-7 |
RTECSNumber | KN4730200 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.