Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl propargyl ether, 98%

Catalog No. p-7035753
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 16314-18-4 | C7H12O | 112.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16314-18-4
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD07368651
InChI Key QIGSPXPODKLGRK-UHFFFAOYSA-N
PubChem CID 7172302
IUPAC Name 2-methyl-2-prop-2-ynoxypropane
SMILES CC(C)(C)OCC#C

Specifications

Density 0.835
Boiling Point 119°C
Flash Point 20°C (68°F)
Refractive Index 1.418
Linear Formula HC≡CCH2OC(CH3)3
Quantity 5 g
UN Number UN1993
Synonym tert-butyl propargyl ether, 3-tert-butoxy prop-1-yne, 1-propyne,3-1,1-dimethylethoxy, 1-propyne, 3-1,1-dimethylethoxy, 3-tert-butoxyprop-1-yne, 3-tert-butyloxy-1-propyne, tert-butylpropargylether
Formula Weight 112.17
Percent Purity 98%
Chemical Name or Material tert-Butyl propargyl ether
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.