Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butylamine, 98%

Catalog No. AAA111350F
Change view
Click to view available options
Quantity:
10,000 mL
1000 mL
2500 mL
500 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA111350F 2500 mL
AAA11135AP 500 mL
AAA11135AU 1000 mL
AAA111350D 10,000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA111350F Supplier Thermo Scientific Chemicals Supplier No. A111350F
Only null left
Add to Cart

CAS: 75-64-9 | C4H11N | 73.139 g/mol

tert-butylamine is used as a precursor for the preparation of rubber accelerators, pharmaceuticals and dyes. It is employed in synthesis of N-tert-butyl-acetamide, isoquinolines, pyridines, decumbenine, sulfontes, sulfonamides and methyl beta-ketophosphonate. It is also used in the study the effect of amines in influencing atmospheric sulfuric acid - water nucleation, hydroamination of terminal alkynes and measuring the surface acidity of solid catalyst. It acts as an indicator molecule in the study of NMR -based pH determination method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-butylamine is used as a precursor for the preparation of rubber accelerators, pharmaceuticals and dyes. It is employed in synthesis of N-tert-butyl-acetamide, isoquinolines, pyridines, decumbenine, sulfontes, sulfonamides and methyl beta-ketophosphonate. It is also used in the study the effect of amines in influencing atmospheric sulfuric acid - water nucleation, hydroamination of terminal alkynes and measuring the surface acidity of solid catalyst. It acts as an indicator molecule in the study of NMR -based pH determination method.

Solubility
Miscible with water, alcohol, ether, chloroform and organic solvents.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acids and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-64-9
Molecular Formula C4H11N
MDL Number MFCD00008050
InChI Key YBRBMKDOPFTVDT-UHFFFAOYSA-N
Synonym tert-butylamine, erbumine, 2-amino-2-methylpropane, t-butylamine, 2-propanamine, 2-methyl, trimethylaminomethane, 1,1-dimethylethylamine, 2-aminoisobutane, 1,1-dimethylethanamine, butylamine, tert
PubChem CID 6385
ChEBI CHEBI:44639
IUPAC Name 2-methylpropan-2-amine
SMILES CC(C)(C)N

Specifications

Melting Point -67°C
Density 0.696
Boiling Point 44°C to 46°C
Flash Point −38°C (−36°F)
Odor Amine-like
Refractive Index 1.378
Quantity 2500 mL
UN Number UN2733
Beilstein 605267
Sensitivity Air Sensitive
Merck Index 14,1545
Solubility Information Miscible with water,alcohol,ether,chloroform and organic solvents.
Molecular Weight (g/mol) 73.139
Formula Weight 73.14
Percent Purity 98%
Chemical Name or Material tert-Butylamine
Show More Show Less
Hazard Category H225-H302-H314-H331-H335-H373
Hazard Statement GHS H Statement
H225-H300-H314-H318-H332
Highly flammable liquid and vapor.
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: I
EINECSNumber 200-888-1
RTECSNumber EO3330000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.