Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyldimethylsilyl trifluoromethanesulfonate, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
69739-34-0
C7H15F3O3SSi
264.33
MFCD00000405
WLLIXJBWWFGEHT-UHFFFAOYSA-N
tert-butyldimethylsilyl trifluoromethanesulfonate,trifluoromethanesulfonic acid tert-butyldimethylsilyl ester,tbdms triflate,t-butyldimethylsilyl triflate,tert-butyldimethylsilyl trifluoromethanesulphonate,tbdms-otf,tert-butyldimethylsilyl triflate,silane tbm2 triflate,t-butyldimethylsilyltrifluoromethanesulfonate,t-butylmethylsilyl triflate
123564
[tert-butyl(dimethyl)silyl] trifluoromethanesulfonate
CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F
69739-34-0
C7H15F3O3SSi
264.33
MFCD00000405
WLLIXJBWWFGEHT-UHFFFAOYSA-N
tert-butyldimethylsilyl trifluoromethanesulfonate,trifluoromethanesulfonic acid tert-butyldimethylsilyl ester,tbdms triflate,t-butyldimethylsilyl triflate,tert-butyldimethylsilyl trifluoromethanesulphonate,tbdms-otf,tert-butyldimethylsilyl triflate,silane tbm2 triflate,t-butyldimethylsilyltrifluoromethanesulfonate,t-butylmethylsilyl triflate
123564
[tert-butyl(dimethyl)silyl] trifluoromethanesulfonate
CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F
Boiling Point 65°C to 67°C (12.0mmHg)
Density 1.1500g/mL
Flash Point 36°C
Specific Gravity 1.15
Refractive Index 1.3850 to 1.388
Quantity 5mL
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Linear Formula CF3SO3Si(CH3)2C(CH3)3
Fieser 10,63; 11,90; 12,86; 13,50; 17,55
Solubility Information Solubility in water: decomposes
Formula Weight 264.33
Physical Form Fuming Liquid
Percent Purity 98%
Chemical Name or Material tert-Butyldimethylsilyl trifluoromethanesulfonate
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 274-102-