Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyldimethylsilyl trifluoromethanesulfonate, 98%

Catalog No. p-3735887
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle

CAS: 69739-34-0 | C7H15F3O3SSi | 264.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69739-34-0
Molecular Formula C7H15F3O3SSi
Molecular Weight (g/mol) 264.33
MDL Number MFCD00000405
InChI Key WLLIXJBWWFGEHT-UHFFFAOYSA-N
Synonym tert-butyldimethylsilyl trifluoromethanesulfonate, trifluoromethanesulfonic acid tert-butyldimethylsilyl ester, tbdms triflate, t-butyldimethylsilyl triflate, tert-butyldimethylsilyl trifluoromethanesulphonate, tbdms-otf, tert-butyldimethylsilyl triflate, silane tbm2 triflate, t-butyldimethylsilyltrifluoromethanesulfonate, t-butylmethylsilyl triflate
PubChem CID 123564
IUPAC Name [tert-butyl(dimethyl)silyl] trifluoromethanesulfonate
SMILES CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F

Specifications

Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 65°C to 67°C (12.0 mmHg)
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Packaging Glass bottle
Refractive Index 1.3850 to 1.388
Linear Formula CF3SO3Si(CH3)2C(CH3)3
Quantity 5 mL
Fieser 10,63; 11,90; 12,86; 13,50; 17,55
Specific Gravity 1.15
Solubility Information Solubility in water: decomposes
Formula Weight 264.33
Percent Purity 98%
Physical Form Fuming Liquid
Chemical Name or Material tert-Butyldimethylsilyl trifluoromethanesulfonate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 274-102-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.